Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Chloropropiophenone

Catalog No. 33561410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-561-410G 10 g
33-561-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-561-410G Supplier Sigma Aldrich Supplier No. 33561410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Chloropropiophenone

Specifications

CAS 936-59-4
Melting Point 48°C to 50°C (lit.)
Boiling Point 113°C to 115°C (4 mmHg)
Molecular Formula C9H9ClO
Linear Formula ClCH2CH2COC6H5
MDL Number MFCD00000990
Quantity 10 g
Synonym beta-Chloropropiophenone; 3-Chloro-1-phenyl-1-propanone
Molecular Weight (g/mol) 168.62
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.