Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyl Chloroformate 95.0+%, TCI America™
SDP

Catalog No. C198325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C198325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C198325G Supplier TCI America Supplier No. C198325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 628-11-5
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 156.99
InChI Key MTXMEFUEBCFWCY-UHFFFAOYSA-N
Synonym Chloroformic Acid 3-Chloropropyl Ester
PubChem CID 69399
IUPAC Name 3-chloropropyl carbonochloridate
SMILES C(COC(=O)Cl)CCl

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 g
UN Number 3277
Formula Weight 156.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.