Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Chloropropyl)diphenylsulfonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. C13801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13801G Supplier TCI America Supplier No. C13801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33462-80-5
Molecular Formula C15H16BClF4S
Molecular Weight (g/mol) 350.607
MDL Number MFCD00074932
InChI Key AVYMGNNILZYVOV-UHFFFAOYSA-N
PubChem CID 2736650
IUPAC Name 3-chloropropyl(diphenyl)sulfanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[S+](CCCCl)C2=CC=CC=C2

Specifications

Melting Point 109°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 350.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (3-Chloropropyl)diphenylsulfonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.