Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyltrichlorosilane 97.0+%, TCI America™
SDP

Catalog No. C084425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C084425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C084425G Supplier TCI America Supplier No. C084425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2550-06-3
Molecular Formula C3H6Cl4Si
Molecular Weight (g/mol) 211.97
MDL Number MFCD00013694
InChI Key OOXSLJBUMMHDKW-UHFFFAOYSA-N
Synonym 3-chloropropyltrichlorosilane, trichloro 3-chloropropyl silane, 3-chloropropyl trichlorosilane, chloropropyltrichlorosilane, silane, trichloro 3-chloropropyl, silane, 3-chloropropyltrichloro, trichloro-3-chloro-propyl-silane, silane, trichloro chloropropyl, g-chloropropyltrichlorosilane, gamma-chloropropyl trichlorosilane
PubChem CID 75693
IUPAC Name trichloro(3-chloropropyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)CCl

Specifications

Color Yellow
Boiling Point 178°C
Quantity 25 g
UN Number 2987
Formula Weight 211.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropyltrichlorosilane
EINECSNumber (2)-2046
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.