Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Chloropropyl)trimethoxysilane

Catalog No. 4401831L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-018-31L 1 L
44-018-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-018-31L Supplier Sigma Aldrich Supplier No. 4401831L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Chloropropyl)trimethoxysilane (CPTMS) is an organo-silane which forms a self-assembled monolayer (SAMs) that facilitates the surface modification of different bio-and nano- materials.CPTMS can be used to silanize a variety of nanoparticles like carbon nanotubes (CNT) and silica nanoparticles (SiO2) for different applications like development for molecularly imprinted polymers for protein detection and fabrication of mesoporous silica(organic-inorganic hybrid). It may be used to functionalize silica-iron oxide nanoparticles which can be used as catalysts for the synthesis of pyranodichromene-6,8-dione derivatives.

Specifications

CAS 2530-87-2
Density 1.09 g/mL (at 25°C)
Boiling Point 195°C (750 mmHg)
Molecular Formula C6H15ClO3Si
Refractive Index n20/D 1.419 (literature)
Linear Formula Cl(CH2)3 Si(OCH3)3
Quantity 1 L
MDL Number MFCD00000997
Synonym delta-Chloropropyltrimethoxysilane; (gamma-Chloropropyl)trimethoxysilane; (3-Chloropropyl)trimethoxysilane; 3-(Trimethoxysilyl)propyl chloride
Molecular Weight (g/mol) 198.72
Percent Purity ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.