Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropyridine, Spectrum™ Chemical
SDP

Catalog No. 18620450 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-450 100 g
18-620-449 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-450 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C2919100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 626-60-8

Specifications

Boiling Point 148°C/298°F
Quantity 100 g
Molecular Weight (g/mol) 113.54
Percent Purity 95%
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material 3-Chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.