Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorothioanisole 96.0+%, TCI America™
SDP

Catalog No. C20705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20705G 5 g
C207025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20705G Supplier TCI America Supplier No. C20705G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4867-37-2
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00040127
InChI Key PTGSDZVASWKUHK-UHFFFAOYSA-N
Synonym 3-chlorothioanisole, 3-chlorophenyl methyl sulfide, 3-chloro thioanisole, 1-chloro-3-methylsulfanyl benzene, m-chlorothioanisole, 3-methylthio chlorobenzene, 3-cpms, 3-chlorothioanisol, pubchem10644, acmc-1bnc1
PubChem CID 97585
IUPAC Name 1-chloro-3-methylsulfanylbenzene
SMILES CSC1=CC(=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 244°C
Quantity 5 g
Formula Weight 158.64
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorothioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.