Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorothiophene-2-carbonyl chloride, Thermo Scientific™

Catalog No. GK01307DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GK01307DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GK01307DA Supplier Thermo Scientific Chemicals Supplier No. GK01307DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86427-02-3
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00068137
InChI Key GCPHKTQMABHWPY-UHFFFAOYSA-N
Synonym 3-chlorothiophene-2-carbonylchloride, 3-chloro-thiophene-2-carbonyl chloride, 3-chloro-2-chlorocarbonyl thiophene, 2-thiophenecarbonyl chloride, 3-chloro-9ci, pubchem5480, timtec-bb sbb005473, buttpark 29\06-44, chloro thiophene-2-carbonyl chloride, 3-chlorothiophene-2-carboyl chloride, 3-chloro-2-thiophenecarbonyl chloride
PubChem CID 2735824
IUPAC Name 3-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=C(Cl)C=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 3-Chlorothiophene-2-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.