Learn More
Cayman Chemical 3β-Cholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502941215
Encompass_Preferred
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS
Catalog No. 50-294-1215 Supplier Cayman Chemical Supplier No. 273761 MG
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS