Learn More
Cayman Chemical 3-CholIc AcId-d4 500ug
SDP
Catalog No. 502937786
Encompass_Preferred
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS
Catalog No. 50-293-7786 Supplier Cayman Chemical Supplier No. 27376500 UG
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS