Learn More
Cayman Chemical 3β-Cholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 5 mg
SDP
Catalog No. 502940997
Encompass_Preferred
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS
Catalog No. 50-294-0997 Supplier Cayman Chemical Supplier No. 273765 MG
An internal standard for the quantification of 3β-cholic acid by GC- or LC-MS