Learn More
AdipoGen 3-cis-Hydroxyglibenclamide
SDP
Catalog No. 502560789
Encompass_Preferred
Chemical. CAS 23074-02-4. Formula C23H28ClN3O6S. MW 510. Synthetic. M2-Metabolite of the antidiabetic compound glyburide. Antidiabetic reagent. Shows 50% hypoglycaemic activity of the parent compound, by increasing insulin secretion. Compound can be used as analytical reference material.
Catalog No. 50-256-0789 Supplier AdipoGen Supplier No. CDXH0030M001
Chemical. CAS 23074-02-4. Formula C23H28ClN3O6S. MW 510. Synthetic. M2-Metabolite of the antidiabetic compound glyburide. Antidiabetic reagent. Shows 50% hypoglycaemic activity of the parent compound, by increasing insulin secretion. Compound can be used as analytical reference material.