Learn More
Medchemexpress LLC 3-Cl-Pyridine-amide-acrylaldehyde-piperazine | 99.3% | 294.74 | 50 MG
SDP
Catalog No. 5000387148
Encompass_Preferred
3-Cl-Pyridine-amide-acrylaldehyde-piperazine is a synthetic intermediate of LO-4-25. LO-4-25 is an androgen receptor (AR) DNA binding domain ligand linked via a linker to the truncated fumaramide handle. This product shows robust degradation of both AR and AR-V7 in 22Rv1 cells.
  • Synthetic intermediate.
  • Degrades androgen receptor (AR) and AR-V7 in 22Rv1 cells.
  • For research use only.
  • Relevant to cancer research.
Catalog No. 50-003-87148 Supplier Medchemexpress LLC Supplier No. HY17606850MG
3-Cl-Pyridine-amide-acrylaldehyde-piperazine is a synthetic intermediate of LO-4-25. LO-4-25 is an androgen receptor (AR) DNA binding domain ligand linked via a linker to the truncated fumaramide handle. This product shows robust degradation of both AR and AR-V7 in 22Rv1 cells.
  • Synthetic intermediate.
  • Degrades androgen receptor (AR) and AR-V7 in 22Rv1 cells.
  • For research use only.
  • Relevant to cancer research.