Learn More
Medchemexpress LLC 3-Cl-Pyridine-amide-acrylaldehyde-piperazine | 99.30% | 294.74 | 100 MG
SDP
Catalog No. 5000387457
Encompass_Preferred
3-Cl-Pyridine-amide-acrylaldehyde-piperazine is a synthetic intermediate of LO-4-25. LO-4-25 is an androgen receptor (AR) DNA binding domain ligand linked via a linker to the truncated fumaramide handle. LO-4-25 shows robust degradation of both AR and AR-V7 in 22Rv1 cells.
  • Synthetic intermediate of LO-4-25
  • Androgen receptor (AR) DNA binding domain ligand
  • Shows robust degradation of both AR and AR-V7 in 22Rv1 cells
  • For research use only
Catalog No. 50-003-87457 Supplier Medchemexpress LLC Supplier No. HY176068100MG
3-Cl-Pyridine-amide-acrylaldehyde-piperazine is a synthetic intermediate of LO-4-25. LO-4-25 is an androgen receptor (AR) DNA binding domain ligand linked via a linker to the truncated fumaramide handle. LO-4-25 shows robust degradation of both AR and AR-V7 in 22Rv1 cells.
  • Synthetic intermediate of LO-4-25
  • Androgen receptor (AR) DNA binding domain ligand
  • Shows robust degradation of both AR and AR-V7 in 22Rv1 cells
  • For research use only