Learn More
NEST SCIENTIFIC INC 3-CODE 2ML CRYOVIAL EXTERNAL
Catalog No. NC2887722
Encompass_Preferred
Three-code-in-one (digital code, barcode, and QR-like code)Vials havelaser-etched international standard DATAMATRIX 2D code on the bottom for traceaility. Vials also have barcodes physically laser etched into their side. Barcodes are indelible, they cannot be scratched off or removed by solvents like DMSO. Cryogenic vials are made from USP grade VI virgin polypropylene and feature leakproof caps, making them suitable for use in liquid phase of LN2. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC2887722 Supplier NEST SCIENTIFIC INC Supplier No. 612841
Three-code-in-one (digital code, barcode, and QR-like code)Vials havelaser-etched international standard DATAMATRIX 2D code on the bottom for traceaility. Vials also have barcodes physically laser etched into their side. Barcodes are indelible, they cannot be scratched off or removed by solvents like DMSO. Cryogenic vials are made from USP grade VI virgin polypropylene and feature leakproof caps, making them suitable for use in liquid phase of LN2. Molded in an ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.