Learn More
Waters Corp Waters Rebuild Kit, 3 Position Select Valve, 2795 Separations Module
Catalog No. 50792382 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Rebuilding the 2790/2795 Three-Column Select Valve involves changing the rotor seal and stator face assembly.
  • Designed for HPLC systems.
  • Includes components for rotor seal and stator face assembly changes.
  • Replaces parts as required.
  • Provided as one unit per package.
Catalog No. 50-792-382 Supplier Waters Corp Supplier No. 700000431
Rebuilding the 2790/2795 Three-Column Select Valve involves changing the rotor seal and stator face assembly.
  • Designed for HPLC systems.
  • Includes components for rotor seal and stator face assembly changes.
  • Replaces parts as required.
  • Provided as one unit per package.