Learn More
Waters Corp Waters Valve, 3 Column Select, 8-port, 3-position
Catalog No. 50792383 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3-Column Selection Valve is an 8-port, 3-position valve that allows selection among 3 column positions. The tubing for the 3-column selection valve is installed at the factory. It can switch between columns, bypass to the detector using a small restrictor, or bypass to waste.
  • 8-port, 3-position valve
  • Allows selection among 3 column positions
  • Tubing is installed at the factory
  • Can switch between columns
  • Can bypass to the detector using a small restrictor
  • Can bypass to waste
Catalog No. 50-792-383 Supplier Waters Corp Supplier No. 700000432
Add to Cart
Add to Cart
Request a Quote
The 3-Column Selection Valve is an 8-port, 3-position valve that allows selection among 3 column positions. The tubing for the 3-column selection valve is installed at the factory. It can switch between columns, bypass to the detector using a small restrictor, or bypass to waste.
  • 8-port, 3-position valve
  • Allows selection among 3 column positions
  • Tubing is installed at the factory
  • Can switch between columns
  • Can bypass to the detector using a small restrictor
  • Can bypass to waste