Learn More
SPECTRUM QUALITY STANDARDS AR1000 [3 Component w/ISTD] | 10x2mL
Supplier:  SPECTRUM QUALITY STANDARDS AR1000
Encompass_Preferred
Check standard consists of 3 components with a Deuterated Internal Standard mixture consisting of Benzene d6, Naphthalene-d8, & Ethylbenzene d10 in a 10×2 ml ampoule set.
Catalog No. NC1576271