Learn More
Med Vet International 3 Corner Beaker, 50ml, J0168, 10 pack
Catalog No. 5000585797
Encompass_Preferred
These graduated beakers are unbreakable polypropylene with built-in pour spouts. Not affected by most chemicals. Beakers are inexpensive enough to be disposable but can be autoclaved and/or reused. 10/pk.
Catalog No. 50-005-85797 Supplier Med Vet International Supplier No. JOR168
These graduated beakers are unbreakable polypropylene with built-in pour spouts. Not affected by most chemicals. Beakers are inexpensive enough to be disposable but can be autoclaved and/or reused. 10/pk.