Learn More
Ted Pella Inc Applicator Sticks, Cotton Tipped, 7.6cm (3"), 1000/cs
SDP
Catalog No. NC0804848
Encompass
Hardwood, cotton-tipped on one end. Available in 7.6cm (3") and 15.2cm (6") length. Can be used for cleaning delicate parts together with our Polishing Compounds. Sold in boxes of 1000.
Catalog No. NC0804848 Supplier Ted Pella Inc Supplier No. 80903
Hardwood, cotton-tipped on one end. Available in 7.6cm (3") and 15.2cm (6") length. Can be used for cleaning delicate parts together with our Polishing Compounds. Sold in boxes of 1000.