Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-2-pyridone, 98%

Catalog No. AAH6425406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6425406 5 g
AAH6425414 25 g
AAH6425422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6425406 Supplier Thermo Scientific Chemicals Supplier No. H6425406

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 20577-27-9 | C6H4N2O | 120.111 g/mol

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20577-27-9
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
MDL Number MFCD00661865
InChI Key DYUMBFTYRJMAFK-UHFFFAOYSA-N
Synonym 3-cyano-2-hydroxypyridine, 2-hydroxynicotinonitrile, 2-hydroxy-3-cyanopyridine, 2-hydroxypyridine-3-carbonitrile, 1,2-dihydro-2-oxopyridine-3-carbonitrile, 2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-cyano-2 1h-pyridinone, 3-cyano-2-pyridone, 3-pyridinecarbonitrile, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-pyridinecarbonitrile
PubChem CID 577543
IUPAC Name 2-oxo-1H-pyridine-3-carbonitrile
SMILES C1=CNC(=O)C(=C1)C#N

Specifications

Melting Point 224°C to 226°C
Quantity 5 g
UN Number UN3439
Beilstein 4661670
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 120.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Cyano-2-pyridone
Hazard Category H302-H318
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.