Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C26761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26761G Supplier TCI America Supplier No. C26761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 214210-21-6
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03095130
InChI Key OLKIYJDSLMKNLC-UHFFFAOYSA-N
Synonym 3-cyano-4-fluorobenzeneboronic acid, 3-cyano-4-fluorophenyl boronic acid, 3-cyano-4-fluoro-phenyl boronic acid, 3-cyano-4-fluorophneylboronic acid, boronic acid, 3-cyano-4-fluorophenyl, 3-cyano-4-fluorobenzeneboronicacid, pubchem1802, acmc-1cq96, ksc494o9l, 3-cyano-4-fluoro phenylboronic acid
PubChem CID 2757964
IUPAC Name (3-cyano-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C#N)(O)O

Specifications

Color White
Quantity 1 g
Formula Weight 164.93
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.