Learn More
Medchemexpress LLC 3-Cyano-5-[2-[4-[2-(3-methylindol-1-yl)ethyl]piperazin-1-yl]phenyl]-N-(3-pyrrolidin-1-ylpropyl)benzamide | 1793053-37-8 | MFCD28902312 | 98.5% | 574.76 g/mol | C36H42N6O | 10 MG
SDP
Catalog No. 5000294658
Encompass_Preferred
LLY-507 is a research-grade small-molecule inhibitor of the protein-lysine methyltransferase SMYD2, used as a chemical probe in epigenetics and cancer biology to study SMYD2-mediated methylation and downstream cellular effects.
  • Potent SMYD2 inhibition (IC50 <15 nM).
  • High purity (98.5%).
  • Chemical formula C36H42N6O; molecular weight 574.76 g/mol.
  • Available in small-molecule quantities, including 10 mg vials.
  • Supplied as solid with recommended DMSO solvent or as a pre-made DMSO solution.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-94658 Supplier Medchemexpress LLC Supplier No. HY1931310MG
LLY-507 is a research-grade small-molecule inhibitor of the protein-lysine methyltransferase SMYD2, used as a chemical probe in epigenetics and cancer biology to study SMYD2-mediated methylation and downstream cellular effects.
  • Potent SMYD2 inhibition (IC50 <15 nM).
  • High purity (98.5%).
  • Chemical formula C36H42N6O; molecular weight 574.76 g/mol.
  • Available in small-molecule quantities, including 10 mg vials.
  • Supplied as solid with recommended DMSO solvent or as a pre-made DMSO solution.
  • For research use only; not for human or veterinary use.