Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-5-fluorophenylboronic acid, 97%, Thermo Scientific™

Catalog No. AC430460010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430460010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430460010 Supplier Thermo Scientific Chemicals Supplier No. 430460010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 304858-67-1
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD09263608
InChI Key DLYWCECHXBOCAS-UHFFFAOYSA-N
Synonym 3-cyano-5-fluorophenyl boronic acid, 3-cyano-5-fluorobenzeneboronic acid, 3-borono-5-fluorobenzonitrile, boronic acid, 3-cyano-5-fluorophenyl-9ci, boronic acid, 3-cyano-5-fluorophenyl, pubchem17351, acmc-1csdr, ksc495g6d, 3-cyano-5-fluorophenylboronicacid
PubChem CID 23435605
IUPAC Name (3-cyano-5-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(=CC(F)=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.93
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 3-Cyano-5-fluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.