Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-5-(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH6126603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6126603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6126603 Supplier Thermo Scientific Chemicals Supplier No. H6126603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyano-5-(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1212021-62-9
Molecular Formula C8H5BF3NO2
Molecular Weight (g/mol) 214.94
MDL Number MFCD03095102
InChI Key CTJAWCWPRYNDDQ-UHFFFAOYSA-N
Synonym 3-cyano-5-trifluoromethyl phenyl boronic acid, 3-cyano-5-trifluoromethyl phenylboronic acid, acmc-209a9l, 3-cyano-5-trifluoromethyl phenyl boronicacid
PubChem CID 56776695
IUPAC Name [3-cyano-5-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC(=C1)C(F)(F)F)C#N

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 214.94
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Cyano-5-(trifluoromethyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.