Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Cyano-7-ethoxycoumarin | 117620-77-6 | ≥98.0% | 5 MG
SDP

Catalog No. 5000410321
Encompass_Preferred

3-Cyano-7-ethoxycoumarin is a fluorogenic cytochrome P-450 substrate that, upon enzyme cleavage, generates a blue fluorescent product. It serves as a fluorescent probe useful in microsomal dealkylase studies and is intended for research use only. This high-purity chemical aids in understanding typical drug-drug interactions resulting from enzyme inhibition.

  • Generates blue fluorescent product upon enzyme cleavage
  • Useful in microsomal dealkylase studies
  • High purity and quality chemical
  • Aids in understanding typical drug-drug interactions
  • Molecular weight of 215.20
  • Chemical formula C12H9NO3
  • Appears as a light yellow to brown solid
  • Emission wavelength of 456
  • Excitation wavelength of 355

Catalog No. 50-004-10321 Supplier Medchemexpress LLC Supplier No. HYD00555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Cyano-7-ethoxycoumarin is a fluorogenic cytochrome P-450 substrate that, upon enzyme cleavage, generates a blue fluorescent product. It serves as a fluorescent probe useful in microsomal dealkylase studies and is intended for research use only. This high-purity chemical aids in understanding typical drug-drug interactions resulting from enzyme inhibition.

  • Generates blue fluorescent product upon enzyme cleavage
  • Useful in microsomal dealkylase studies
  • High purity and quality chemical
  • Aids in understanding typical drug-drug interactions
  • Molecular weight of 215.20
  • Chemical formula C12H9NO3
  • Appears as a light yellow to brown solid
  • Emission wavelength of 456
  • Excitation wavelength of 355

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.