Learn More
AdipoGen 3-Cyano-7-ethoxycoumarin
SDP
Supplier:  AdipoGen CDXC0092M020
Encompass_Preferred
Chemical. CAS 117620-77-6. Formula C12H9NO3. MW 215.2. Synthetic. Substrate for determination of cytochrom P-450 dependent oxidases about 50-100 times more sensitive than conventional substrate ethoxy-resorufin due to higher turn over rate.
Catalog No. 50-256-1481