Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. TL00666EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00666EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00666EA Supplier Thermo Scientific Chemicals Supplier No. TL00666EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56542-67-7
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
MDL Number MFCD00203480
InChI Key BHNRGBRMCNHNQD-UHFFFAOYSA-N
Synonym 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463
PubChem CID 2801360
IUPAC Name 3-cyanobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CNC6H4SO2Cl
Quantity 10 g
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzene-1-sulfonyl chloride
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.