Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. C15105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15105G 5 g
C15101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15105G Supplier TCI America Supplier No. C15105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 874-97-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD01321087
InChI Key PCOFIIVWHXIDGT-UHFFFAOYSA-N
Synonym 3-cyanobenzyl alcohol, 3-hydroxymethyl benzonitrile, 3-cyanobenzylalcohol, benzonitrile, 3-hydroxymethyl, 3-hyroxymethylbenzonitrile, 3-hydroxymethyl benzenecarbonitrile, m-cyanobenzyl alcohol, 3-cyanophenyl methanol, acmc-209qo5
PubChem CID 2734677
IUPAC Name 3-(hydroxymethyl)benzonitrile
SMILES C1=CC(=CC(=C1)CO)C#N

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyanobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.