Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanobenzylsulfonyl chloride, 97%, Thermo Scientific™

Catalog No. MO07947DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07947DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07947DA Supplier Thermo Scientific Chemicals Supplier No. MO07947DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56106-01-5
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.651
MDL Number MFCD09046692
InChI Key YNWYDZBBHOWDAE-UHFFFAOYSA-N
Synonym 3-cyanophenyl methanesulfonyl chloride, 3-cyanobenzylsulfonyl chloride, 3-cyanophenyl methanesulphonyl chloride, 3-chlorosulphonyl methyl benzonitrile, 3-cyano-phenyl-methanesulfonyl chloride, 3-cyanobenzylsulphonyl chloride, 3-cyano benzylsulfonyl chloride, benzenemethanesulfonylchloride, 3-cyano, 3-cyano-phenyl methanesulfonyl chloride, 3-chlorosulfonyl methyl benzenecarbonitrile
PubChem CID 16774314
IUPAC Name (3-cyanophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)CS(=O)(=O)Cl)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.66
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzylsulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.