Learn More
Medchemexpress LLC 3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid | 83249-02-9 | MFCD19229172 | 98.8% | 137.14 g/mol | C7H7NO2 | 250 MG
SDP
Catalog No. 5000209195
Encompass_Preferred
3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid is a chemical intermediate used in the synthesis of active pharmaceutical compounds. It is supplied in small research quantities and is accompanied by a certificate of analysis (COA) and a safety data sheet (SDS).
  • Drug intermediate used in the synthesis of active pharmaceutical compounds.
  • High purity: 98.8%.
  • Available in small-scale research sizes: 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.
  • Certificate of analysis and safety data sheet provided.
  • Suitable for research and development applications.
Catalog No. 50-002-09195 Supplier Medchemexpress LLC Supplier No. 250MGHYW065054
3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid is a chemical intermediate used in the synthesis of active pharmaceutical compounds. It is supplied in small research quantities and is accompanied by a certificate of analysis (COA) and a safety data sheet (SDS).
  • Drug intermediate used in the synthesis of active pharmaceutical compounds.
  • High purity: 98.8%.
  • Available in small-scale research sizes: 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.
  • Certificate of analysis and safety data sheet provided.
  • Suitable for research and development applications.