Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanocoumarin, 97%, Thermo Scientific™

Catalog No. AC456590050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456590050 Supplier Thermo Scientific Chemicals Supplier No. 456590050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15119-34-3
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00115699
InChI Key QKJALQPLNMEDAV-UHFFFAOYSA-N
Synonym 3-cyanocoumarin, 2-oxo-2h-chromene-3-carbonitrile, coumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 2-oxo, 2-oxo-2h-1-benzopyran-3-carbonitrile, coumarin, 3-cyano-7ci,8ci, 3-cyano-coumarin, pubchem8675, acmc-1cfdq
PubChem CID 203763
IUPAC Name 2-oxochromene-3-carbonitrile
SMILES O=C1OC2=CC=CC=C2C=C1C#N

Specifications

Melting Point 184.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 171.15
Percent Purity 97%
Chemical Name or Material 3-Cyanocoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.