Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Cyanopropyltriethoxysilane

Catalog No. 37415625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-415-625G 25 g
37-415-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-415-625G Supplier Sigma Aldrich Supplier No. 37415625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-cyanopropyltriethoxysilane (CPTS) is a trialkoxysilane with a reactive cyanoalkyl group attached to the silicon atom.CPTS is used in the synthesis of titanium based sorbent for solid phase extraction(SPE) of aromatic amines from river water. It finds its application in the development of organically modified silicate films coated on gold electrodes for the detection of ametryn. It may also be used to modify the surface characteristics of biopolymers by using polar functionalities for the analysis of non-steroidal anti-inflammatory drugs(NSAIDs) in river water.

Specifications

CAS 1067-47-6
Density 0.966 g/mL (at 25°C)
Boiling Point 100°C to 101°C (6 mmHg)
Molecular Formula C10H21NO3Si
Refractive Index n20/D 1.417 (literature)
Linear Formula (C2H5O)3 Si(CH2)3 CN
Quantity 25 g
MDL Number MFCD00042636
Synonym 4-(Triethoxysilyl)butyronitrile
Molecular Weight (g/mol) 231.36
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.