Learn More
Medchemexpress LLC 3-Cyanoumbelliferone | 19088-73-4 | ≥98.0% | 250 MG
SDP
Catalog No. 5000414346
Encompass_Preferred
3-Cyanoumbelliferone is a coumarin derivative that acts as a molecular probe and fluorescent dye. It is also a macrophage migration inhibitory factor (MIF) tautomerase inhibitor with a Ki of 2.9 μM.
  • Acts as a molecular probe and fluorescent dye.
  • Macrophage migration inhibitory factor (MIF) tautomerase inhibitor.
  • Has a molecular weight of 187.15.
  • Chemical formula is C10H5NO3.
  • Appears as a solid with a yellow to orange color.
  • Has a solubility of 6.67 mg/mL in DMSO (35.64 mM), requiring ultrasonic, warming, and heating to 60 °C.
Catalog No. 50-004-14346 Supplier Medchemexpress LLC Supplier No. 250MGHYD0039
Add to Cart
Add to Cart
3-Cyanoumbelliferone is a coumarin derivative that acts as a molecular probe and fluorescent dye. It is also a macrophage migration inhibitory factor (MIF) tautomerase inhibitor with a Ki of 2.9 μM.
  • Acts as a molecular probe and fluorescent dye.
  • Macrophage migration inhibitory factor (MIF) tautomerase inhibitor.
  • Has a molecular weight of 187.15.
  • Chemical formula is C10H5NO3.
  • Appears as a solid with a yellow to orange color.
  • Has a solubility of 6.67 mg/mL in DMSO (35.64 mM), requiring ultrasonic, warming, and heating to 60 °C.