Learn More
AdipoGen 3-Cyanoumbelliferone
SDP
Supplier:  AdipoGen CDXC0007G001
Encompass_Preferred
Chemical. CAS 19088-73-4. Formula C10H5NO3. MW 187.15. Synthetic. Phenolic dyes with high pKas - such as 7-hydroxycoumarin often called umbelliferone and the more common 7-hydroxy-4-methylcoumarin - are not fully deprotonated and therefore not fully fluorescent unless the pH of the reaction mixture is raised to above pH ~10. Thus, substrates derived from these fluorophores are seldom used for continuous measurement of enzymatic activity in solution or live cells. The similar 3-cyano-7-hydroxycoumarin and 6,8-difluoro-7-hydroxy-4-methylcoumarin have lower pKas, making them suitable for a broader range of applications. Moreover, the -trifluoromethylumbelliferone product has longer-wavelength spectra and stronger absorbance than -methylumbelliferone. Ether, ester and phosphate substrates derived from these phenolic dyes may be fluorescent but invariably exhibit shorter-wavelength absorption and emission spectra.
Catalog No. 50-256-1562