Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-cyclohexen-1-ol, 4-methyl-1-(1-methylethyl)- | 562-74-3 | MFCD00001562 | 98.6% | 154.25 | C10H18O | 1 G
SDP

Catalog No. 5000300601
Encompass_Preferred

Terpinen-4-ol is a naturally occurring monoterpene alcohol used as an analytical standard and research reagent. It is supplied as a colorless to light yellow liquid, and is characterized by a documented GC assay and certificate of analysis for quality control.

  • Naturally occurring monoterpene alcohol.
  • Colorless to light yellow liquid.
  • Molecular formula C10H18O; molecular weight 154.25 g/mol.
  • High GC purity (98.6%).
  • Store at 4°C and protect from light; in solvent: -80°C for long-term storage.

Catalog No. 50-003-00601 Supplier Medchemexpress LLC Supplier No. HYW0173161G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Terpinen-4-ol is a naturally occurring monoterpene alcohol used as an analytical standard and research reagent. It is supplied as a colorless to light yellow liquid, and is characterized by a documented GC assay and certificate of analysis for quality control.

  • Naturally occurring monoterpene alcohol.
  • Colorless to light yellow liquid.
  • Molecular formula C10H18O; molecular weight 154.25 g/mol.
  • High GC purity (98.6%).
  • Store at 4°C and protect from light; in solvent: -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.