Learn More
Medchemexpress LLC 3-cyclohexen-1-ol, 4-methyl-1-(1-methylethyl)- | 562-74-3 | MFCD00001562 | 98.6% | 154.25 g/mol | C10H18O | 50 MG
SDP
Catalog No. 5000299554
Encompass_Preferred
Terpinen-4-ol is a naturally occurring monoterpene used as a research reagent and reference standard. It exhibits reported bactericidal, antifungal, anti-inflammatory, and antitumor activities and is commonly used in biochemical, pharmacological, and analytical studies requiring a reliable monoterpene standard.
  • High purity (98.6% by GC) suitable for analytical use.
  • Suitable as a reference standard for GC method development and quantification.
  • Soluble in common organic solvents for ease of sample preparation.
  • Naturally occurring monoterpene; chemical formula C10H18O; molecular weight 154.25 g/mol.
  • Supplied with batch-specific certificate of analysis for traceability.
Catalog No. 50-002-99554 Supplier Medchemexpress LLC Supplier No. HYW017316R50MG
Terpinen-4-ol is a naturally occurring monoterpene used as a research reagent and reference standard. It exhibits reported bactericidal, antifungal, anti-inflammatory, and antitumor activities and is commonly used in biochemical, pharmacological, and analytical studies requiring a reliable monoterpene standard.
  • High purity (98.6% by GC) suitable for analytical use.
  • Suitable as a reference standard for GC method development and quantification.
  • Soluble in common organic solvents for ease of sample preparation.
  • Naturally occurring monoterpene; chemical formula C10H18O; molecular weight 154.25 g/mol.
  • Supplied with batch-specific certificate of analysis for traceability.