Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-cyclohexen-1-ol, 4-methyl-1-(1-methylethyl)- | 562-74-3 | MFCD00001562; MFCD00167108 | 98.7% | 154.25 g/mol | C10H18O | 250 MG
SDP

Catalog No. 5000303732
Encompass_Preferred

Terpinen-4-ol is a naturally occurring monoterpene alcohol used in biochemical and pharmacological research for its reported bactericidal, antifungal, anti-inflammatory, and antitumor activities. Supplied as a characterized liquid with supporting analytical data, it is intended for research applications that require a documented natural product.

  • High purity: 98.65% (GC) as reported on COA.
  • Physical form: colorless to light yellow liquid.
  • Analytical support: 1H NMR and optical rotation provided on COA.
  • Storage guidance: protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • Applications: suitable for antimicrobial, antifungal, and anti-inflammatory research.

Catalog No. 50-003-03732 Supplier Medchemexpress LLC Supplier No. 250MGHYW017316
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Terpinen-4-ol is a naturally occurring monoterpene alcohol used in biochemical and pharmacological research for its reported bactericidal, antifungal, anti-inflammatory, and antitumor activities. Supplied as a characterized liquid with supporting analytical data, it is intended for research applications that require a documented natural product.

  • High purity: 98.65% (GC) as reported on COA.
  • Physical form: colorless to light yellow liquid.
  • Analytical support: 1H NMR and optical rotation provided on COA.
  • Storage guidance: protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • Applications: suitable for antimicrobial, antifungal, and anti-inflammatory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.