Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-cyclohexene-1-carboxaldehyde, 4-(4-hydroxy-4-methylpentyl)- | 31906-04-4 | MFCD00019423 | 97.8% | 210.31 | C13H22O2 | 250 MG
SDP

Catalog No. 5000303059
Encompass_Preferred

Hydroxyisohexyl 3-cyclohexene carboxaldehyde is an aldehyde fragrance ingredient used as a hapten probe and analytical reference in research settings. Supplied as a colorless to light-yellow liquid, it is employed in contact allergy studies, patch-testing research, and analytical method development.

  • Liquid, colorless to light yellow appearance.
  • Molecular formula C13H22O2; molecular weight 210.31.
  • Purity 97.8% suitable for analytical and research use.
  • Available in multiple pack sizes to accommodate laboratory needs.
  • Serves as a hapten probe in immunological and contact allergy investigations.
  • Appropriate for reference standard and method development applications.

Catalog No. 50-003-03059 Supplier Medchemexpress LLC Supplier No. 250MGHYW035214
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxyisohexyl 3-cyclohexene carboxaldehyde is an aldehyde fragrance ingredient used as a hapten probe and analytical reference in research settings. Supplied as a colorless to light-yellow liquid, it is employed in contact allergy studies, patch-testing research, and analytical method development.

  • Liquid, colorless to light yellow appearance.
  • Molecular formula C13H22O2; molecular weight 210.31.
  • Purity 97.8% suitable for analytical and research use.
  • Available in multiple pack sizes to accommodate laboratory needs.
  • Serves as a hapten probe in immunological and contact allergy investigations.
  • Appropriate for reference standard and method development applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.