Learn More
Medchemexpress LLC 3-cyclohexene-1-carboxaldehyde, 4-(4-hydroxy-4-methylpentyl)- | 31906-04-4 | MFCD00019423 | 97.8% | 210.31 g/mol | C13H22O2 | 100 MG
SDP
Catalog No. 5000299850
Encompass_Preferred
Hydroxyisohexyl 3-cyclohexene carboxaldehyde is a synthetic fragrance aldehyde used as a hapten probe in contact-allergy and fragrance research. Supplied as a colorless to light yellow liquid with high analytical purity, it is available neat or as preformed solutions in DMSO for biological assays. Store under inert atmosphere at low temperature to maintain stability.
Catalog No. 50-002-99850 Supplier Medchemexpress LLC Supplier No. HYW035214100MG
Hydroxyisohexyl 3-cyclohexene carboxaldehyde is a synthetic fragrance aldehyde used as a hapten probe in contact-allergy and fragrance research. Supplied as a colorless to light yellow liquid with high analytical purity, it is available neat or as preformed solutions in DMSO for biological assays. Store under inert atmosphere at low temperature to maintain stability.