Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Cyclohexene-1-methanol

Catalog No. 162167100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-216-7100G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-216-7100G Supplier Sigma Aldrich Supplier No. 162167100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Cyclohexene-1-methanol

Specifications

CAS 1679-51-2
Density 0.961 g/mL (at 25°C (literature))
Boiling Point 190°C to 192°C (lit.), 80°C to 85°C (18 mmHg)
Molecular Formula C7H12O
Refractive Index n20/D 1.484 (literature)
Linear Formula C6H9CH2OH
MDL Number MFCD00001574
Quantity 500 mg
Synonym 1,2,3,6-Tetrahydrobenzyl alcohol; Cyclohexen-4-ylmethanol
Molecular Weight (g/mol) 112.17
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.