Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-96334
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294390010 1 g Glass bottle
AC294390100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294390010 Supplier Thermo Scientific Chemicals Supplier No. 294390010
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4771-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
MDL Number MFCD00013781
InChI Key VUSWCWPCANWBFG-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
PubChem CID 20903
IUPAC Name cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 17.0°C
Color Blue-Gray to Brown-Yellow
Density 1.0800g/mL
Boiling Point 238.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Quantity 1 g
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 126.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Cyclohexenecarboxylic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
EINECSNumber 225-314-7
RTECSNumber GW3675000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.