Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexyl-1-propanol, 99%

Catalog No. AC255880050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255880050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255880050 Supplier Thermo Scientific Chemicals Supplier No. 255880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1124-63-6 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-63-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00001528
InChI Key CLYAQFSQLQTVNO-UHFFFAOYSA-N
Synonym cyclohexanepropanol, 3-cyclohexyl-1-propanol, 3-cyclohexylpropanol, unii-kjb3vo906f, kjb3vo906f, cyclohexylpropyl alcohol, 3-hydroxypropylcyclohexane, 3-cyclohexylpropyl alcohol, acmc-1c5ti
PubChem CID 70763
IUPAC Name 3-cyclohexylpropan-1-ol
SMILES C1CCC(CC1)CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60°C
Color Colorless
Density 0.9100g/mL
Boiling Point 223°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4650 to 1.467
Linear Formula C6H11CH2CH2CH2OH
Quantity 5 mL
Specific Gravity 0.91
Solubility Information Solubility in water: 0.06% (20 c)
Viscosity 36.7 mPa.s (20°C)
Formula Weight 142.24
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 3-Cyclohexyl-1-propanol, 99%
Show More Show Less
EINECSNumber 214-401-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.