Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexylaminopropanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. C0921250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0921250G 250 g
C092125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0921250G Supplier TCI America Supplier No. C0921250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym caps, 3-cyclohexylaminopropanesulfonic acid, n-cyclohexyl-3-aminopropanesulfonic acid, 3-cyclohexylamino-1-propanesulfonic acid, 1-propanesulfonic acid, 3-cyclohexylamino, 3-cyclohexylamino propane-1-sulfonic acid, unii-4w981o1lxp, 3-cyclohexylamino propanesulfonic acid, 3-cyclohexylaminopropane-1-sulphonic acid, 3-cyclohexyl-1-propylsulfonic acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Color White
Quantity 250 g
Formula Weight 221.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyclohexylaminopropanesulfonic Acid [Good′s buffer component for biological research]
RTECSNumber TZ6395000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.