Learn More
Accela Chembio Inc MF: C6H8O2 | MW: 112.13 | >97% | BP: 227.3 celsius at 760 mmHg | Density: 1.084.
Catalog No. 50353867
Encompass_Preferred
MF: C6H8O2 | MW: 112.13 | >97% | BP: 227.3 celsius at 760 mmHg | Density: 1.084.
Catalog No. 50-353-867 Supplier Accela Chembio Inc Supplier No. SY00381325G
MF: C6H8O2 | MW: 112.13 | >97% | BP: 227.3 celsius at 760 mmHg | Density: 1.084.