Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)-N-isopropylbenzamide | 1917347-65-9 | 99.8% | 299.31 g/mol | C15H19F2NO3 | 10 MG
SDP

Catalog No. 5000227803
Encompass_Preferred

FCPR03 is a small-molecule selective phosphodiesterase 4 (PDE4) inhibitor supplied for research use. It is a solid research compound with high reported purity and is commonly used in in vitro studies of inflammation and neurobiology.

  • Selective PDE4 inhibitor activity
  • High reported purity (≈99.8%)
  • Solid, white to off-white appearance
  • Soluble in DMSO: 100 mg/mL; ready for reconstitution as 10 mM in DMSO
  • Molecular weight approximately 299.31 g/mol
  • Available in multiple small pack sizes for research use

Catalog No. 50-002-27803 Supplier Medchemexpress LLC Supplier No. HY11797710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FCPR03 is a small-molecule selective phosphodiesterase 4 (PDE4) inhibitor supplied for research use. It is a solid research compound with high reported purity and is commonly used in in vitro studies of inflammation and neurobiology.

  • Selective PDE4 inhibitor activity
  • High reported purity (≈99.8%)
  • Solid, white to off-white appearance
  • Soluble in DMSO: 100 mg/mL; ready for reconstitution as 10 mM in DMSO
  • Molecular weight approximately 299.31 g/mol
  • Available in multiple small pack sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.