Learn More
Medchemexpress LLC 3-(cyclopropylmethoxy)-4-(difluoromethoxy)-N-isopropylbenzamide | 1917347-65-9 | 99.8% | 299.31 g·mol⁻¹ | C15H19F2NO3 | 100 MG
SDP
Catalog No. 5000226789
Encompass_Preferred
FCPR03 is a small-molecule research compound that functions as a potent and selective phosphodiesterase 4 (PDE4) inhibitor. It is provided for laboratory research use and supplied in solid and solution formats for experimental flexibility.
  • Potent, selective PDE4 inhibitor with reported low-nanomolar IC50 values.
  • High purity (~99.8%) suitable for in vitro studies.
  • Molecular weight 299.31 g·mol⁻¹; molecular formula C15H19F2NO3.
  • Available as a 100 mg solid and as 10 mM DMSO solutions for convenient dosing.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-26789 Supplier Medchemexpress LLC Supplier No. HY117977100MG
FCPR03 is a small-molecule research compound that functions as a potent and selective phosphodiesterase 4 (PDE4) inhibitor. It is provided for laboratory research use and supplied in solid and solution formats for experimental flexibility.
  • Potent, selective PDE4 inhibitor with reported low-nanomolar IC50 values.
  • High purity (~99.8%) suitable for in vitro studies.
  • Molecular weight 299.31 g·mol⁻¹; molecular formula C15H19F2NO3.
  • Available as a 100 mg solid and as 10 mM DMSO solutions for convenient dosing.
  • Intended for research use only; not for human or clinical use.