Learn More
Medchemexpress LLC 3-(cyclopropylmethoxy)-4-(difluoromethoxy)-N-isopropylbenzamide | 1917347-65-9 | 99.8% | 299.31 g/mol | C15H19F2NO3 | 25 MG
SDP
Catalog No. 5000227820
Encompass_Preferred
A potent, selective phosphodiesterase 4 (PDE4) inhibitor used as a research compound for in vitro and preclinical pharmacology. Reported IC50 values include catalytic domain 60 nM, PDE4B1 31 nM, and PDE4D7 47 nM. The compound shows strong selectivity over other PDE families and is supplied at high reported purity.
  • Potent and selective PDE4 inhibition.
  • Low-nanomolar IC50 values for multiple PDE4 isoforms.
  • High selectivity over other phosphodiesterase families.
  • Suitable for in vitro pharmacology and preclinical studies.
  • Supplied with high reported purity for reproducible results.
Catalog No. 50-002-27820 Supplier Medchemexpress LLC Supplier No. HY11797725MG
A potent, selective phosphodiesterase 4 (PDE4) inhibitor used as a research compound for in vitro and preclinical pharmacology. Reported IC50 values include catalytic domain 60 nM, PDE4B1 31 nM, and PDE4D7 47 nM. The compound shows strong selectivity over other PDE families and is supplied at high reported purity.
  • Potent and selective PDE4 inhibition.
  • Low-nanomolar IC50 values for multiple PDE4 isoforms.
  • High selectivity over other phosphodiesterase families.
  • Suitable for in vitro pharmacology and preclinical studies.
  • Supplied with high reported purity for reproducible results.