Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(cyclopropylmethyl)-7-[(4-phenyl-1-piperidinyl)methyl]-8-(trifluoromethyl)-triazolo[4,3-a]pyridine | 1254980-38-5 | 98.8% | 50 MG
SDP

Catalog No. 5000231709
Encompass_Preferred

JNJ-46281222 is a metabotropic glutamate receptor 2 (mGlu2) selective positive allosteric modulator (PAM) used as a research reagent for in vitro pharmacology and receptor modulation studies.

  • Selective mGlu2 positive allosteric modulator with nanomolar affinity (Kd = 1.7 nM)
  • High modulatory potency (pEC50 = 7.71)
  • Molecular formula C23H25F3N4 and molecular weight 414.47 g·mol⁻¹
  • Purity approximately 98.8%
  • Solid, white to off-white appearance
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)
  • For research use only; not for human or clinical use

Catalog No. 50-002-31709 Supplier Medchemexpress LLC Supplier No. HY12053050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JNJ-46281222 is a metabotropic glutamate receptor 2 (mGlu2) selective positive allosteric modulator (PAM) used as a research reagent for in vitro pharmacology and receptor modulation studies.

  • Selective mGlu2 positive allosteric modulator with nanomolar affinity (Kd = 1.7 nM)
  • High modulatory potency (pEC50 = 7.71)
  • Molecular formula C23H25F3N4 and molecular weight 414.47 g·mol⁻¹
  • Purity approximately 98.8%
  • Solid, white to off-white appearance
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)
  • For research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.