Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(cyclopropylmethyl)-7-((4-phenylpiperidin-1-yl)methyl)-8-(trifluoromethyl)-triazolopyridine | 1254980-38-5 | 98.8% | 5 MG
SDP

Catalog No. 5000234392
Encompass_Preferred

JNJ-46281222 is an mGlu2-selective positive allosteric modulator used as a research tool in receptor binding and functional assays; it has nanomolar affinity and is supplied as a high-purity solid or DMSO solution for in vitro studies.

  • Metabotropic glutamate receptor 2 (mGlu2)-selective positive allosteric modulator
  • High affinity: Kd = 1.7 nM
  • Functional potency: pEC50 = 7.71 (EC50 ≈ 19-21 nM in CHO assays)
  • Molecular formula C23H25F3N4; molecular weight 414.47 g·mol-1
  • High purity (98.77%)
  • Available as solid powder and as 10 mM solution in DMSO
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • For research use only; not for human or clinical use

Catalog No. 50-002-34392 Supplier Medchemexpress LLC Supplier No. HY1205305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JNJ-46281222 is an mGlu2-selective positive allosteric modulator used as a research tool in receptor binding and functional assays; it has nanomolar affinity and is supplied as a high-purity solid or DMSO solution for in vitro studies.

  • Metabotropic glutamate receptor 2 (mGlu2)-selective positive allosteric modulator
  • High affinity: Kd = 1.7 nM
  • Functional potency: pEC50 = 7.71 (EC50 ≈ 19-21 nM in CHO assays)
  • Molecular formula C23H25F3N4; molecular weight 414.47 g·mol-1
  • High purity (98.77%)
  • Available as solid powder and as 10 mM solution in DMSO
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • For research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.